Anti-DMXL2 Rabbit Polyclonal Antibody

Supplier: Proteintech
24415-1-AP-150UL
10085-578EA 402 USD
10085-578
Anti-DMXL2 Rabbit Polyclonal Antibody
Antibodies
DMXL2, also named as rabconnectin-3, is a functional regulator of mammalian notch signaling with WD domains. Rabconnectin-3 is abundantly expressed in the brain where it is enriched in the synaptic vesicle fraction. Immunofluorescence and immunoelectron microscopy revealed that rabconnectin-3 was concentrated on synaptic vesicles at synapses. It may serve as a scaffold molecule for both Rab3 GEP and GAP on synaptic vesicles. In western blotting, we got 340 kDa and 104 kDa in mouse tissues. The two bands may represent different isoforms.

Western Blot: Mouse Brain Tissue, 1:200-1:1000; IHC: Human cerebellum Tissue, 1:20-1:200

Type: Primary
Antigen: DMXL2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR