Anti-Barkor Rabbit Polyclonal Antibody

Supplier: Proteintech
24412-1-AP-150UL
10083-508EA 402 USD
10083-508
Anti-Barkor Rabbit Polyclonal Antibody
Antibodies
Barkor, also named as ATG14, Atg14L and KIAA0831, is named as Beclin 1-associated autophagy-related key regulator protein. The function of Barkor in autophagy has been manifested in several assays, including stress-induced LC3 lipidation, autophagosome formation, and Salmonella typhimurium amplification.

Western Blot: Mouse Brain Tissue, 1:200-1:2000; IF: HeLa Cells, 1:10-1:100; IHC: Human Brain Tissue, 1:20-1:200

Type: Primary
Antigen: Barkor
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR