Anti-KTELC1 Rabbit Polyclonal Antibody

Supplier: Proteintech
24409-1-AP-150UL
10088-742EA 402 USD
10088-742
Anti-KTELC1 Rabbit Polyclonal Antibody
Antibodies
KTELC1 also termed as CLP46 is a glucosyltransferase, which implicates in Protein modification and glycosylation. KTELC1 encodes a 392 amino acids protein and its molecular weight is 46kda, but is exists glycosylation modification. KTELC1 is localized in endoplasmic reticulum lumen and abundantly expressed in liver, brain, heart, skeletal muscle, spleen, kidney, placenta, lung and peripheral blood leukocyte. KTELC1 may regulation in cell proliferation and Notch signal pathway.

Western Blot: HepG2 Cells, 1:200-1:2000; IF: HepG2 Cells, 1:10-1:100; IHC: Human Liver Tissue, 1:20-1:200

Type: Primary
Antigen: KTELC1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR