Anti-GTF2E1 Rabbit Polyclonal Antibody

Supplier: Proteintech
24405-1-AP-150UL
10087-954EA 402 USD
10087-954
Anti-GTF2E1 Rabbit Polyclonal Antibody
Antibodies
Western Blot: HEK-293 Cells, 1:1000-1:10000

Type: Primary
Antigen: GTF2E1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR