Anti-MLK4 Rabbit Polyclonal Antibody

Supplier: Proteintech
24395-1-AP-150UL
10090-274EA 402 USD
10090-274
Anti-MLK4 Rabbit Polyclonal Antibody
Antibodies
MLK4 belongs to the MAP kinase kinase kinase subfamily. The MLK family members are characterized by the presence of signature sequences ofSer/Thr and Tyr kinases within their catalytic domain. The MLK1-4 contains an aminoterminal Src-Homology (SH3) domain, followed by the kinase domain, a leucine-zipper region, and a Cdc42/Rac-interactive binding (CRIB) motif. MLK4 expressed in two splicing variants, MLK4α and MLK4β. The difference of these two isoforms is located in their C-terminal portion. The negative effect of MLK4 on LPS-induced ERK and JNK activation could be very important in controlling the level of cytokine expression, and thus maintaining the balance of inflammatory responses.. This protein has 3 isoforms produced by alternative splicing with the molecular weight of 63 kDa, 113 kDa, 53 kDa.

Western Blot: HeLa Cells, 1:500-1:5000; IF:C6 Cells, 1:10-1:100; IHC: Human Brain Tissue, 1:20-1:200

Type: Primary
Antigen: MLK4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR