Anti-C17orf85 Rabbit Polyclonal Antibody

Supplier: Proteintech
24384-1-AP-150UL
10083-302EA 402 USD
10083-302
Anti-C17orf85 Rabbit Polyclonal Antibody
Antibodies
chromosome 17 open reading frame 85(C17orf85)also named as Protein ELG is 620 amino acids protein, which is localizes in human chromosome 17. The function of protein is still non-known and it consists of some phosphorylation site . It may involves in human embryonic stem cell differentiation.

Western Blot: HeLa Cells, 1:500-1:5000; IF: HeLa Cells, 1:10-1:100

Type: Primary
Antigen: C17orf85
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR