Anti-ACTN3 Rabbit Polyclonal Antibody

Supplier: Proteintech
24378-1-AP-150UL
10081-236EA 402 USD
10081-236
Anti-ACTN3 Rabbit Polyclonal Antibody
Antibodies
Western Blot: Mouse Skeletal Muscle Tissue, 1:200-1:1000

Type: Primary
Antigen: ACTN3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR