Anti-TMEM25 Rabbit Polyclonal Antibody

Supplier: Proteintech
24361-1-AP-150UL
10095-686EA 402 USD
10095-686
Anti-TMEM25 Rabbit Polyclonal Antibody
Antibodies
TMEM25 is a novel member of the immunoglobulin superfamily. BC042896 and AY358919 cDNAs are derived from human TMEM25 gene. TMEM25 isoform 1 (BC042896), consisting of exons 1-9, encodes a 366-aa transmembrane protein. TMEM25 isoform 2 (AY358919), consisting of exons 1-4 and 6-9, encodes a 322-aa secreted protein . Human TMEM25 gene are found to encode transmembrane-type as well as secreted-type proteins due to alternative splicing of exon-skipping type. TMEM25 mRNA is expressed in brain, including cerebellar cortex and hippocampus, as well as in neuroblastoma, brain tumors, and gastric cancer. TMEM25 is a target of pharmacogenomics in the field of oncology and regenerative medicine. TMEM25 mRNAs may be useful members of a panel of favorable prognostic and predictive markers for breast cancer( PMID:19776672).

Western Blot: Mouse Liver Tissue, 1:500-1:5000; IHC: Human Stomach cancer Tissue, 1:20-1:200

Type: Primary
Antigen: TMEM25
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR