Anti-IFT43 Rabbit Polyclonal Antibody

Supplier: Proteintech
24338-1-AP-150UL
10083-766EA 402 USD
10083-766
Anti-IFT43 Rabbit Polyclonal Antibody
Antibodies
IFT43, encoded by C14ORF179, is a subunit of the intraflagellar transport complex A (IFT-A) of primary cilia. The IFT-A is implicated in retrograde ciliary transport along axonemal microtubules. Defects in subunits of IFT-A has been associated with skeletal ciliopathies, including Sensenbrenner syndrome, Jeune syndrome, and Ellis-van-Creveld syndrome. Mutations in C14ORF179 has been linked to Sensenbrenner syndrome. (21378380)

IHC: Human Testis Tissue, 1:20-1:200

Type: Primary
Antigen: IFT43
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR