Anti-ATG16L2 Rabbit Polyclonal Antibody

Supplier: Proteintech
24322-1-AP-150UL
10083-240EA 402 USD
10083-240
Anti-ATG16L2 Rabbit Polyclonal Antibody
Antibodies
ATG16L2 also named as Autophagy related protein 16-2 is a 619 amino-acid protein, Which belongs to the WD repeat ATG16 family. ATG16L2 contains 7 WD repeats and localizes in the cytoplasm. There are three isoforms of ATG16L2 and the molecular weight of three isoform is respectively 69kDa, 56kDa and 31kDa. ATG16L2 may play a role in autophagy.

Western Blot: Mouse spleen Tissue, 1:500-1:5000; IP: Mouse Testis Tissue, 1:200-1:2000

Type: Primary
Antigen: ATG16L2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR