Anti-DGKB Rabbit Polyclonal Antibody

Supplier: Proteintech
24320-1-AP-150UL
10085-792EA 402 USD
10085-792
Anti-DGKB Rabbit Polyclonal Antibody
Antibodies
DGKB(Diacylglycerol kinase beta) is also named as DAGK2, KIAA0718 , 90 kDa diacylglycerol kinase and belongs to the eukaryotic diacetylglycerol kinase family. It is a key enzyme in lipid metabolism that functions to reintroduce diacylglycerol formed from the hydrolysis of phospholipids into the biosynthetic pathway. This protein has 2 isoforms produced by alternative splicing with the molecular weight of 87 kDa and 91 kDa.

Western Blot: HeLa Cells, 1:500-1:5000

Type: Primary
Antigen: DGKB
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR