Anti-LYSMD3 Rabbit Polyclonal Antibody

Supplier: Proteintech
24313-1-AP-150UL
10089-440EA 402 USD
10089-440
Anti-LYSMD3 Rabbit Polyclonal Antibody
Antibodies
LYSMD3 also termed as LysM and putative peptidoglycan binding domain containing protein 3 is a 306 amino acid single-pass membrane protein that contains one LysM repeat. LysM domain encoding genes may play a role in brain and in nervous system development and functioning. LYSMD3, shows a high level of central nervous expression during embryogenesis and is also, therefore, a good candidate gene for other central nervous system (CNS) symptoms, such as psychomotor retardation, brain anomalies and muscular hypotonia of the 5q14.3 microdeletion syndrome.

Western Blot:Neuro-2a Cells, 1:200-1:1000

Type: Primary
Antigen: LYSMD3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR