Anti-KDM2A Rabbit Polyclonal Antibody

Supplier: Proteintech
24311-1-AP-150UL
10088-216EA 402 USD
10088-216
Anti-KDM2A Rabbit Polyclonal Antibody
Antibodies
KDM2A(Lysine-specific demethylase 2A) is also named as CXXC8, FBL7, FBXL11, JHDM1A, KIAA1004 and belongs to the JHDM1 histone demethylase family. It is required to maintain the heterochromatic state, and playing a role in epigenetic silencing. KDM2A is one of the four subunits of the E3 ubiquitin protein ligase complex (SCF) with SKP1, CUL1, RBX1, Fbox protein bringing ubiquitin conjugating enzymes to substrates recruited by F-box.

Western Blot:Jurkat Cells, 1:500-1:5000

Type: Primary
Antigen: KDM2A
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR