Anti-EEPD1 Rabbit Polyclonal Antibody

Supplier: Proteintech
24310-1-AP-150UL
10086-284EA 402 USD
10086-284
Anti-EEPD1 Rabbit Polyclonal Antibody
Antibodies
EEPD1 is also known as KIAA1706, is a 198 amino-acid protein, Which belongs to endonuclease/exonuclease/phosphatase family domain containing 11. EEPD1 is localized in the cell nucleus and may involve in the splicing of DNA. The function of EEPD1 is still non-known and need to be further studied.

Western Blot:COLO 320 Cells, 1:200-1:2000

Type: Primary
Antigen: EEPD1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR