Anti-LARGE Rabbit Polyclonal Antibody

Supplier: Proteintech
24307-1-AP-150UL
10089-514EA 402 USD
10089-514
Anti-LARGE Rabbit Polyclonal Antibody
Antibodies
Western Blot: Mouse Kidney Tissue, 1:500-1:5000

Type: Primary
Antigen: LARGE
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR