Anti-peptide YY Rabbit Polyclonal Antibody

Supplier: Proteintech
24294-1-AP-150UL
10092-088EA 402 USD
10092-088
Anti-peptide YY Rabbit Polyclonal Antibody
Antibodies
IHC: Human pancreas Tissue, 1:20-1:200

Type: Primary
Antigen: peptide YY
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR