Anti-RFTN1 Rabbit Polyclonal Antibody

Supplier: Proteintech
24289-1-AP-150UL
10093-598EA 402 USD
10093-598
Anti-RFTN1 Rabbit Polyclonal Antibody
Antibodies
IHC: Human tonsillitis Tissue, 1:20-1:200; Western Blot: HeLa Cells, 1:1000-1:10000

Type: Primary
Antigen: RFTN1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR