Anti-ANKRD53 Rabbit Polyclonal Antibody

Supplier: Proteintech
24283-1-AP-150UL
10082-908EA 402 USD
10082-908
Anti-ANKRD53 Rabbit Polyclonal Antibody
Antibodies
ANKRD53 also termed as ankyrin repeat domain 53 is a 533 amino-acid protein,which contains three ANK repeats and belongs to ankyrin family. The function of ANKRD53 is still non-known and need to be further studied. Ankyrins are membrane adaptor molecules that play important roles in coupling integral membrane proteins to the spectrin-based cytoskeleton network.

Western Blot: HeLa Cells, 1:500-1:5000; IHC: Human Kidney Tissue, 1:20-1:200

Type: Primary
Antigen: ANKRD53
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR