Anti-MEFV Rabbit Polyclonal Antibody

Supplier: Proteintech
24280-1-AP-150UL
10090-118EA 402 USD
10090-118
Anti-MEFV Rabbit Polyclonal Antibody
Antibodies
Western Blot: Human plasma Tissue, 1:500-1:5000

Type: Primary
Antigen: MEFV
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR