Anti-CCDC104 Rabbit Polyclonal Antibody

Supplier: Proteintech
24276-1-AP-150UL
10084-160EA 402 USD
10084-160
Anti-CCDC104 Rabbit Polyclonal Antibody
Antibodies
The CCDC104(Coiled-coil domain-containing protein 104) protein, whose function is unknown, is expressed in various human tissues, including the brain, and is localized mainly to the nucleus, but is also found in the cytoplasm. It may act as an effector for ARL3. This protein has 2 isoforms produced by alternative splicing.

Western Blot: Mouse lung Tissue, 1:500-1:5000; IHC: Human Testis Tissue, 1:20-1:200; IF:A431 Cells, 1:10-1:100

Type: Primary
Antigen: CCDC104
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR