Anti-C1ORF31 Rabbit Polyclonal Antibody

Supplier: Proteintech
24209-1-AP-150UL
10083-804EA 402 USD
10083-804
Anti-C1ORF31 Rabbit Polyclonal Antibody
Antibodies
C1orf31, also known as COA6, is a conserved mitochondrial protein required for respiratory complex IV biogenesis. Recently study reported that C1orf31 is required for maintenance of cytochrome c oxidase (CcO) subunit levels, and has role in the Cu delivery pathway to CcO. Defects in C1orf31 is associated with mitochondrial respiratory chain disease (MRCD). (24549041)

Western Blot: HEK-293 Cells, 1:500-1:5000; IP: HEK-293 Cells, 1:200-1:2000

Type: Primary
Antigen: C1ORF31
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR