Anti-GSK3B Rabbit Polyclonal Antibody

Supplier: Proteintech
24198-1-AP-150UL
10081-826EA 402 USD
10081-826
Anti-GSK3B Rabbit Polyclonal Antibody
Antibodies
Western Blot: Mouse Brain Tissue, 1:500-1:5000; IF:Ethacrynic acid treated HeLa Cells, 1:10-1:100; IHC: Human Testis Tissue, 1:20-1:200; FC: HeLa Cells, N/A

Type: Primary
Antigen: GSK3B
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR