Anti-C2orf67 Rabbit Polyclonal Antibody

Supplier: Proteintech
24183-1-AP-150UL
10083-846EA 402 USD
10083-846
Anti-C2orf67 Rabbit Polyclonal Antibody
Antibodies
C2orf67,also termed as chromosome 2 open reading frame 67, encodes a 705 amino-acid protein. It exists a variant, which encodes 987 amino-acid protein. The C2orf56 may express in lymphoid /immune and nervous system. The specific function of C2orf67 is still non-known.

Western Blot:PC-3 Cells, 1:500-1:5000; IHC: Human Testis Tissue, 1:20-1:200

Type: Primary
Antigen: C2orf67
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR