Anti-JNK1 Rabbit Polyclonal Antibody

Supplier: Proteintech
24164-1-AP-150UL
10081-590EA 537.7 USD
10081-590
Anti-JNK1 Rabbit Polyclonal Antibody
Antibodies
Three members of c-Jun N-terminal kinases (JNK), JNK1, JNK2 and JNK3, have been reported, which are distant members of the MAP kinase family. JNK1 is activated by dual phosphorylation at a Thr-Pro-Tyr motif during response to UV light. JNK1 functions to phosphorylate c-Jun at N-terminal serine regulatory sites of Ser-63 and Ser-73, mapping within the transactivation domain. Phosphorylation of these sites in response to UV results in transcriptional activation of c-Jun.

Western Blot: HEK-293 Cells, 1:500-1:5000; IHC: Human cervical cancer Tissue, 1:20-1:200

Type: Primary
Antigen: JNK1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR