Anti-TMEM59 Rabbit Polyclonal Antibody

Supplier: Proteintech
24134-1-AP-150UL
10095-694EA 402 USD
10095-694
Anti-TMEM59 Rabbit Polyclonal Antibody
Antibodies
IHC: Human breast cancer Tissue, 1:20-1:200; Western Blot: MCF-7 Cells, 1:200-1:1000; IF: HepG2 Cells, 1:10-1:100

Type: Primary
Antigen: TMEM59
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR