Anti-SOD2 Rabbit Polyclonal Antibody

Supplier: Proteintech
24127-1-AP-150UL
10094-994EA 537.7 USD
10094-994
Anti-SOD2 Rabbit Polyclonal Antibody
Antibodies
SOD2(superoxide dismutase 2, mitochondrial ) is also named as IPOB, MNSOD, SODM, Mn-SOD and belongs to the iron/manganese superoxide dismutase family. It is a marker of mitochondria, which is restricted to the perinuclear area in a cell with aggregate formation of mutant SOD1. It functions as the first line of antioxidant defense against highly reactive superoxide radicals and it appears to be early predictors for survival in septic patients with with MIF. It has 2 isoforms with the molecular weight of 25 kDa and 21 kDa.

Western Blot: HeLa Cells, 1:500-1:5000; IF: HUVEC Cells, 1:20-1:200; IHC: Human breast cancer Tissue, 1:20-1:200

Type: Primary
Antigen: SOD2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR