Anti-HACE1 Rabbit Polyclonal Antibody

Supplier: Proteintech
24104-1-AP-150UL
10088-022EA 402 USD
10088-022
Anti-HACE1 Rabbit Polyclonal Antibody
Antibodies
HACE1(E3 ubiquitin-protein ligase HACE1) also named as KIAA1320, has a tumor-suppressor function dependent on its E3 ligase activity and controling adhesion-dependent growth and cell cycle progression during cell stress through degradation of cyclin D1. It belongs to the HECT family of E3 ubiqutin protein ligase. This protein has 4 isoforms produced by alternative splicing with the molecular weight of 102 kDa, 36 kDa, 64 kDa and 78 kDa.

Western Blot: HEK-293 Cells, 1:200-1:2000

Type: Primary
Antigen: HACE1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR