Anti-ANKLE1 Rabbit Polyclonal Antibody

Supplier: Proteintech
24080-1-AP-150UL
10082-894EA 402 USD
10082-894
Anti-ANKLE1 Rabbit Polyclonal Antibody
Antibodies
Western Blot: Hela Cells, 1:200-1:2000; IHC: Human breast cancer Tissue, 1:20-1:200

Type: Primary
Antigen: ANKLE1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR