Anti-RAC1 Rabbit Polyclonal Antibody

Supplier: Proteintech
24072-1-AP-150UL
10081-448EA 402 USD
10081-448
Anti-RAC1 Rabbit Polyclonal Antibody
Antibodies
RAC1b, also named as TC25, belongs to the small GTPase superfamily and Rho family. RAC1 is a plasma membrane-associated small GTPase which cycles between active GTP-bound and inactive GDP-bound states. In its active state, it binds to a variety of effector proteins to regulate cellular responses such as secretory processes, phagocytosis of apoptotic cells, epithelial cell polarization and growth-factor induced formation of membrane ruffles.

Western Blot: Human Brain Tissue, 1:500-1:5000

Type: Primary
Antigen: RAC1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR