Anti-MATN2 Rabbit Polyclonal Antibody

Supplier: Proteintech
24064-1-AP-150UL
10089-998EA 402 USD
10089-998
Anti-MATN2 Rabbit Polyclonal Antibody
Antibodies
The matrilins represent a new family of oligomeric non-collagenous extracellular matrix proteins that are assumed to act as adapter molecules connecting other proteins and proteoglycans in the extracellular matrix . Four matrilins (matrilin-1, -2, -3, and -4) have been identified. They all share a structure made up of von Willebrand factor A domains, epidermal growth factor-like domains and a coiled-coil alpha-helical module . Matrilin-1 and matrilin-3 are expressed mainly in cartilage, while matrilin-2 and matrilin-4 show broader tissue distribution. Matrilin-2 is the largest member of the family. It incorporates into loose and dense connective tissue and becomes associated with some basement membranes .

Western Blot:NIH/3T3 Cells, 1:200-1:2000

Type: Primary
Antigen: MATN2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR