Anti-TMEM27 Rabbit Polyclonal Antibody

Supplier: Proteintech
24059-1-AP-150UL
10081-172EA 402 USD
10081-172
Anti-TMEM27 Rabbit Polyclonal Antibody
Antibodies
TMEM27(Transmembrane protein 27) is also named as collectrin and belongs to the TMEM27 family. It is a pancreatic beta-cell surface protein promoting the preservation of functional beta-cell mass and may be responsible for the sodium retention in salt-sensitive hypertension. This full length has two glycosylation sites.

Western Blot: HEK-293 Cells, 1:200-1:1000

Type: Primary
Antigen: TMEM27
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR