Anti-HSN2 Rabbit Polyclonal Antibody

Supplier: Proteintech
24055-1-AP-150UL
10088-496EA 402 USD
10088-496
Anti-HSN2 Rabbit Polyclonal Antibody
Antibodies
HSN2, also named as WNK1, KDP, KIAA0344, PRKWNK1, P65 and hWNK1, is a serine/threonine kinase which plays an important role in the regulation of electrolyte homeostasis, cell signaling, survival, and proliferation. It acts as an activator and inhibitor of sodium-coupled chloride cotransporters and potassium-coupled chloride cotransporters respectively. WNK1 may also play a role in actin cytoskeletal reorganization. This antibody detects the 65 kDa protein (Erythrocyte 65 kDa protein, P65).

Western Blot: Mouse Brain Tissue, 1:500-1:5000; IHC: Human Kidney Tissue, 1:20-1:200

Type: Primary
Antigen: HSN2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR