Anti-GCR Rabbit Polyclonal Antibody

Supplier: Proteintech
24050-1-AP-150UL
10091-280EA 402 USD
10091-280
Anti-GCR Rabbit Polyclonal Antibody
Antibodies
NR3C1 is a receptor for glucocorticoids, which owns a dual mode of action: as a transcription factor that binds to glucocorticoid response elements (GRE) and as a modulator of other transcription factors. It is involved in cell proliferation and differentiation and specifically implicated in newborn birth weight, thus providing a biological mechanism by which NR3C1 expression may influence birth weight . It also act as a coactivator for STAT5-dependent transcription upon growth hormone (GH) stimulation and could reveal an essential role of hepatic GR in the control of body growth . This antibody is specific to NR3C1.

Western Blot: HeLa Cells, 1:500-1:5000; IF: HepG2 Cells, 1:20-1:200

Type: Primary
Antigen: Glucocorticoid receptor
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR