Anti-CK23 Rabbit Polyclonal Antibody

Supplier: Proteintech
24049-1-AP-150UL
10089-388EA 402 USD
10089-388
Anti-CK23 Rabbit Polyclonal Antibody
Antibodies
Keratins are a large family of proteins that form the intermediate filament cytoskeleton of epithelial cells, which are classified into two major sequence types. Type I keratins are a group of acidic intermediate filament proteins, including K9–K23, and the hair keratins Ha1–Ha8. Type II keratins are the basic or neutral courterparts to the acidic type I keratins, including K1–K8, and the hair keratins, Hb1–Hb6. KRT 23 belongs to type I keratins.

Western Blot:BxPC-3 Cells, 1:500-1:5000; IF: HeLa Cells, 1:10-1:100; IHC: Human Colon cancer Tissue, 1:20-1:200

Type: Primary
Antigen: CK23
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR