Anti-B4GALNT2 Rabbit Polyclonal Antibody

Supplier: Proteintech
24024-1-AP-150UL
10083-460EA 402 USD
10083-460
Anti-B4GALNT2 Rabbit Polyclonal Antibody
Antibodies
B4GALNT2, also named as GALGT2, catalyzes the last step in the biosynthesis of the human Sd(a) antigen, which is found on more than 90% of Caucasian red blood cells. It also catalyzes the last step in the biosynthesis of the Cad antigen, which is related both serologically and biochemically to the Sd(a) antigen. B4GALNT2 transfers a beta-1,4-linked GalNAc to the galactose residue of an alpha-2,3-sialylated chain. This antibody detects the 55-65 kDa B4GALNT2 protein.

Western Blot:Jurkat Cells, 1:500-1:5000; IHC: Human Colon cancer Tissue, 1:20-1:200

Type: Primary
Antigen: B4GALNT2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR