Anti-ANKRD13C Rabbit Polyclonal Antibody

Supplier: Proteintech
24000-1-AP-150UL
10082-902EA 402 USD
10082-902
Anti-ANKRD13C Rabbit Polyclonal Antibody
Antibodies
ANKRD13C, also termed as ankyrin repeat domain 13C, belongs to the ankyrin repeat domain 13 family. ANKRD13C is localized on ER membranes where it binds to DP to control its biogenesis and trafficking. ANKRD13C modulates the expression and maturation of other G pretein coupled receptors as well, indicating that ANKRD13C is a novel regulator of the biogenesis and trafficking of G pretein coupled receptors through the biosynthetic pathway.

Western Blot: MDA-MB-453s Cells, 1:200-1:2000

Type: Primary
Antigen: ANKRD13C
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR