Anti-TFAM Rabbit Polyclonal Antibody

Supplier: Proteintech
23996-1-AP-150UL
10081-164EA 402 USD
10081-164
Anti-TFAM Rabbit Polyclonal Antibody
Antibodies
mtTFA (mitochondrial transcription factor A), also known as mtTF1, TFAM, TCF6 (for transcription factor 6-like1), TCF6L2 and tsHMG, is a nuclear-encoded gene product that is imported into the mitochondria. mtTFA is required for many aspects of mitochondrial biogenesis including the replication and transcription of mitochondrial DNA (mtDNA). In mouse, testis-specific mtTFA is missing the mitochondria targeting sequence and is present in the nucleus rather than the mitochondria. This form of mtTFA is located primarily in the nuclei of elongated spermatids and may be involved in the regulation of gene expression of the haploid male genome. During mouse and human spermatogenesis there is a reduction of mtTFA protein levels and a reduction in mtDNA copy number. These features may provide one of the mechanisms by which paternal mtDNA transmission is prevented. mtTFA has been associated with mitochondrial disorder in humans characterized by ocular myopathy, exercise intolerance and muscle wasting. This antibody specifically recognizes the 25kd human TFAM protein.

  • Western Blot: HEK-293 Cells, 1:500-1:5000; IHC: Human Kidney Tissue, 1:20-1:200

Type: Primary
Antigen: TFAM
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR