Anti-BRMS1L Rabbit Polyclonal Antibody

Supplier: Proteintech
23989-1-AP-150UL
10083-688EA 402 USD
10083-688
Anti-BRMS1L Rabbit Polyclonal Antibody
Antibodies
Western Blot: MCF-7 Cells, 1:200-1:2000

Type: Primary
Antigen: BRMS1L
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR