Anti-GR repeat Rabbit Polyclonal Antibody

Supplier: Proteintech
23978-1-AP-150UL
10087-828EA 537.7 USD
10087-828
Anti-GR repeat Rabbit Polyclonal Antibody
Antibodies
The C9orf72 "GGGGCC" repeat sequence codes five repeat peptide "GA repeat; GAGAGAGAGA", "GP repeat; GPGPGPGPG", "GR repeat; GRGRGRGRG", "AP repeat; APAPAPAPA" and "PR repeat; PRPRPRPRP". This antibody is used to detect the GR repeated sequence. The recombinant proteins GST-SNRPD1 and 6*His tag- SNRPD1 which contains the sequence “GRGRGRGRGRGRGRGRGR” were used to detect this antibody. Both proteins can be recognized by the antibody 23978-1-AP.

Western Blot: Recombinant Protein, 1:500-1:5000

Type: Primary
Antigen: GR repeat
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR