Anti-BARHL2 Rabbit Polyclonal Antibody

Supplier: Proteintech
23976-1-AP-150UL
10083-506EA 402 USD
10083-506
Anti-BARHL2 Rabbit Polyclonal Antibody
Antibodies
BARHL2 also named as BarH like homeobox 2 is 279 amino acid protein, which contains 1 homeobox DNA-binding domain. BARHL2 localizes in nucleus and belongs to BAR homeobox family. BARHL2 as a transcription factor binds optimally to the DNA consensus sequence. Barx2 may differentially control the expression of L1 and other target genes during embryonic development. Barx2 is expressed in several smooth muscle-containing tissues, as well as skeletal muscle, brain, tongue and esophagus.

Western Blot: HepG2 Cells, 1:200-1:1000; IHC: Human Testis Tissue, 1:20-1:200

Type: Primary
Antigen: BARHL2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR