Anti-RCOR2 Rabbit Polyclonal Antibody

Supplier: Proteintech
23969-1-AP-150UL
10093-472EA 402 USD
10093-472
Anti-RCOR2 Rabbit Polyclonal Antibody
Antibodies
In mammals, the CoREST (corepressor for element-1-silencing transcription factor) complex is a chromatin-modifying structure that, through interactions with NRSF (neuron restrictive silencer factor), regulates neuronal gene expression and neuronal cell fate. RCOR2 (REST corepressor 2) and RCOR3 (REST corepressor 3) are nuclear proteins that each contain one ELM2 domain and two SANT domains. RCOR2 and RCOR3, both members of the CoREST family, are thought to function as components of the CoREST complex, possibly playing a role in the transcriptional repression of neuronal genes. Additionally, RCOR2 and RCOR3, in conjunction with CoREST, can form immunocomplexes with a variety of histone-modifying genes, including G9a and HDAC1. Via these protein complexes, RCOR2 and RCOR3 can further regulate transcription by controlling the methylation and demethylation of target genes during early development.

Western Blot:Jurkat Cells, 1:500-1:5000

Type: Primary
Antigen: RCOR2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR