Anti-CLEC14A Rabbit Polyclonal Antibody

Supplier: Proteintech
23964-1-AP-150UL
10084-852EA 524.48 USD
10084-852
Anti-CLEC14A Rabbit Polyclonal Antibody
Antibodies
CLEC14A is a single-pass type I transmembrane protein whose extracellular domain consists of a C-type lectin-like domain (CTLD), a series of epidermal growth factor-like domains, and a sushi-like domain . CLEC14A is a member of the C-type lectin domain superfamily that also includes endosialin, thrombomodulin and CD93 . It has been reported that CLEC14A is specifically expressed in endothelial cells and mediates cell to cell adhesion . CLEC14A may be involved in tumor angiogenesis .

Western Blot: Human Heart Tissue, 1:500-1:5000

Type: Primary
Antigen: CLEC14A
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR