Anti-MAP4K1 Rabbit Polyclonal Antibody

Supplier: Proteintech
23950-1-AP-150UL
10089-942EA 402 USD
10089-942
Anti-MAP4K1 Rabbit Polyclonal Antibody
Antibodies
MAP4K1(Mitogen-activated protein kinase kinase kinase kinase 1) is also named as MEKKK 1, HPK1 and belongs to the protein kinase superfamily. It may function as a novel tumor suppressor and its loss plays a critical role in pancreatic cancer. MAP4K1 regulates stress responses, proliferation, and apoptosis in hematopoietic cells. This protein has 2 isoforms produced by alternative splicing.

Western Blot: Raji Cells, 1:500-1:5000

Type: Primary
Antigen: MAP4K1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR