Anti-GFAP Rabbit Polyclonal Antibody

Supplier: Proteintech
23935-1-AP-150UL
10081-800EA 402 USD
10081-800
Anti-GFAP Rabbit Polyclonal Antibody
Antibodies
GFAP (Glial fibrillary acidic protein ), an intermediate-filament (IF) protein , is specifically expressed in cells of astroglial lineage and is widely used to mark the astroglia in the brain. It is also used as a marker for intracranial and intraspinal tumors arising from astrocytes.

Western Blot: Mouse Brain Tissue, 1:500-1:5000; IHC: Mouse Brain, 1:20-1:200

Type: Primary
Antigen: GFAP
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR