Anti-PPP2R5E Rabbit Polyclonal Antibody

Supplier: Proteintech
23885-1-AP-150UL
10092-724EA 402 USD
10092-724
Anti-PPP2R5E Rabbit Polyclonal Antibody
Antibodies
Western Blot: HeLa Cells, 1:500-1:5000; IHC: Human cervical cancer Tissue, 1:20-1:200

Type: Primary
Antigen: PPP2R5E
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR