Anti-ARCN1 Rabbit Polyclonal Antibody

Supplier: Proteintech
23843-1-AP-150UL
10082-304EA 402 USD
10082-304
Anti-ARCN1 Rabbit Polyclonal Antibody
Antibodies
Archain 1 (ARCN1), also known as delta-COP, is a subunit of the coat protein I (COPI) complex, which comprises one of the protein coats that mediate vesicle budding from the membrane. Protein sequences of ARCN1 are well conserved among eukaryotes and this protein may play a fundamental role in eukaryotic cell biology. It has similarities to heat shock proteins and clathrin-associated proteins, and may be involved in vesicle structure or trafficking. The gene of ARCN1 has been identified in chromosome band 11q23.3, and sequencing of cDNAs suggests that at least two forms of mRNA with alternative 5' ends are present.

  • Western Blot: HepG2 Cells, 1:500-1:5000; IHC: Human Testis Tissue, 1:20-1:200

Type: Primary
Antigen: ARCN1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR