Anti-CTSG Rabbit Polyclonal Antibody

Supplier: Proteintech
23840-1-AP-150UL
10084-104EA 402 USD
10084-104
Anti-CTSG Rabbit Polyclonal Antibody
Antibodies
CTSG(Cathepsin G) is also named as CTLA, CG, MGC23078, Catg and belongs to the peptidase S1 family. It may participate in the killing and digestion of engulfed pathogens, and in connective tissue remodeling at sites of inflammation. CTSG may also play a role in the progression to heart failure by activating angiotensin II, leading to detrimental effects on the heart. The full length protein contains 255 amino acid and has a singal peptide, a propeptide and a glycosylation site.

Western Blot: MCF-7 Cells, 1:200-1:2000

Type: Primary
Antigen: CTSG
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR