Anti-FATE1 Rabbit Polyclonal Antibody

Supplier: Proteintech
23809-1-AP-150UL
10086-882EA 402 USD
10086-882
Anti-FATE1 Rabbit Polyclonal Antibody
Antibodies
FATE1 ,also named as Fetal and adult testis expressed transcript protein, express predominantly in testis, with some expression in lung, heart, kidney, adrenal gland and whole brain. FATE might represent a novel target gene of SF-1 in testicular differentiation and germ cell development. FATE1 lalso involves in the regulation of a wide variety of cellular processes ,including the cell cycle, immune responses, signaling cascades and developmental events through target proteins, such as cyclins, cyclin-dependent kinase inhibitors.

Western Blot: Human Testis Tissue, 1:200-1:2000

Type: Primary
Antigen: FATE1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR