Anti-ZNF271 Rabbit Polyclonal Antibody

Supplier: Proteintech
23799-1-AP-150UL
10097-330EA 402 USD
10097-330
Anti-ZNF271 Rabbit Polyclonal Antibody
Antibodies
ZNF271, also named as ZNF-EB, HZF7, ZNF271-dp, may be involved in transcriptional regulation. It plays a role in the EBV-mediated transformation process. ZNF271 has two isoforms with MW 76 kDa and 48 kDa. This antibody recognize both isoforms.

Western Blot: Mouse pancreas Tissue, 1:500-1:5000

Type: Primary
Antigen: ZNF271
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR