Anti-AKNA Rabbit Polyclonal Antibody

Supplier: Proteintech
23785-1-AP-150UL
10082-720EA 402 USD
10082-720
Anti-AKNA Rabbit Polyclonal Antibody
Antibodies
AKNA also named as AT hook transcription factor is a 1439 amino-acids protein. AKNA, encodes a lymphoid-specific AT-hook transcription factor that is highly expressed by B and T lymphocytes. It coordinately regulates expression of the CD40 receptor and its ligand (CD154), two molecules that are critical for antigen-dependent B-cell development. AKNA expresses at least nine distinct transcripts, some of which are expressed in a tissue-specific manner. These transcripts are products of alternative splicing, alternative promoter usage, and alternative polyadenylation. The alternative AKNA transcripts are predicted to encode overlapping protein isoforms, some of which (p70 and p100)

Western Blot: Raji Cells, 1:200-1:2000; IHC: Human tonsillitis Tissue, 1:20-1:200

Type: Primary
Antigen: AKNA
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR